Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DCLRE1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004002347.4 | XP_004002396.2 | 1 - 2 | 417 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Ovis aries |