Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALMS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027967005.1 | XP_027822806.1 | 22 - 23 | 13231 | 63 | |
XM_027967006.1 | XP_027822807.1 | 22 - 23 | 13180 | 63 | ||
XM_027967007.1 | XP_027822808.1 | 21 - 22 | 13060 | 63 | ||
XM_027967008.1 | XP_027822809.1 | 23 - 24 | 11473 | 63 | ||
XM_027967010.1 | XP_027822811.1 | 23 - 24 | 11233 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Ovis aries |