Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLRN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004020266.4 | XP_004020315.2 | 1 - 2 | 342 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Ovis aries |