Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PM20D2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015097387.2 | XP_014952873.2 | 2 - 3 | 661 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Ovis aries |