Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRAPPC2L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015100411.2 | XP_014955897.1 | 4 - 5 | 431 | 82 | |
XM_027977743.1 | XP_027833544.1 | 4 - 5 | 372 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Ovis aries |