Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BBIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012102918.3 | XP_011958308.1 | 4 - 5 | 302 | 83 | |
XM_012102919.3 | XP_011958309.1 | 3 - 4 | 292 | 83 | ||
XM_012102920.3 | XP_011958310.1 | 3 - 4 | 500 | 83 | ||
XM_012102921.3 | XP_011958311.1 | 3 - 4 | 529 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Ovis aries |