Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC114113228 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027965211.1 | XP_027821012.1 | 1 | 233 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Ovis aries |