Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSPAN19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004006232.4 | XP_004006281.2 | 8 - 9 | 763 | 107 | |
XM_004006233.4 | XP_004006282.2 | 8 - 9 | 733 | 107 | ||
XM_012174038.3 | XP_012029428.2 | 9 - 10 | 847 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Ovis aries |