Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EPGN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004009909.3 | XP_004009958.1 | 4 - 5 | 1277 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Ovis aries |