Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPNE7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027977761.1 | XP_027833562.1 | 13 - 14 | 2210 | 64 | |
XM_027977762.1 | XP_027833563.1 | 14 - 15 | 1538 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Ovis aries |