Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUTF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004015079.4 | XP_004015128.1 | 4 - 5 | 404 | 79 | |
XM_027978101.1 | XP_027833902.1 | 4 - 5 | 451 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Ovis aries |