Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KDELR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004021057.4 | XP_004021106.1 | 1 - 2 | 227 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Ovis aries |