Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAMP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004012943.4 | XP_004012992.1 | 2 - 3 | 221 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Ovis aries |