Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POLG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004013031.3 | XP_004013080.2 | 7 - 8 | 1543 | 95 | |
XM_012186340.2 | XP_012041730.2 | 6 - 7 | 1402 | 95 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Ovis aries |