Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZFP90 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027978116.1 | XP_027833917.1 | 2 - 3 | 115 | 105 | |
XM_027978117.1 | XP_027833918.1 | 2 - 3 | 135 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |