Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LLGL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015098677.2 | XP_014954163.2 | 9 - 10 | 1101 | 98 | |
XM_027974801.1 | XP_027830602.1 | 9 - 10 | 1092 | 98 | ||
XM_027974802.1 | XP_027830603.1 | 9 - 10 | 1089 | 98 | ||
XM_027974803.1 | XP_027830604.1 | 9 - 10 | 1092 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Ovis aries |