Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NXN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027975166.1 | XP_027830967.1 | 3 - 4 | 702 | 101 | |
XM_027975167.1 | XP_027830968.1 | 3 - 4 | 418 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |