Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TEX28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015104973.2 | XP_014960459.2 | 2 - 3 | 794 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Ovis aries |