Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBE2L3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015101751.2 | XP_014957237.1 | 3 - 4 | 382 | 81 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Ovis aries |