Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: YIPF7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012179894.3 | XP_012035284.1 | 5 - 6 | 725 | 57 | |
XM_012179896.3 | XP_012035286.1 | 5 - 6 | 697 | 57 | ||
XM_012179898.3 | XP_012035288.2 | 4 - 5 | 568 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Ovis aries |