Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: THSD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027973586.1 | XP_027829387.1 | 1 - 2 | 1671 | 85 | |
XM_027973587.1 | XP_027829388.1 | 2 - 3 | 273 | 85 | ||
XM_027973588.1 | XP_027829389.1 | 2 - 3 | 328 | 85 | ||
XM_027973589.1 | XP_027829390.1 | 1 - 2 | 1673 | 85 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Ovis aries |