Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C2H2orf80 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027964850.1 | XP_027820651.1 | 6 - 7 | 512 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Ovis aries |