Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USP35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027959278.1 | XP_027815079.1 | 11 - 12 | 3145 | 94 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Ovis aries |