Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACTBL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027980065.1 | XP_027835866.1 | 1 | 323 | 77 | |
XM_027980066.1 | XP_027835867.1 | 2 | 197 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Ovis aries |