Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLSCR5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012101849.3 | XP_011957239.1 | 5 - 6 | 762 | 85 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Ovis aries |