Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KERA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004006250.4 | XP_004006299.1 | 2 - 3 | 1087 | 92 | |
XM_012174075.3 | XP_012029465.1 | 3 - 4 | 1194 | 92 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Ovis aries |