Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARMC7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027974060.1 | XP_027829861.1 | 1 - 2 | 399 | 77 | |
XM_027974061.1 | XP_027829862.1 | 1 - 2 | 403 | 77 | ||
XM_027974062.1 | XP_027829863.1 | 1 - 2 | 396 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Ovis aries |