Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPRD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027974383.1 | XP_027830184.1 | 4 - 5 | 1527 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Ovis aries |