Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C4H7orf57 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004007698.3 | XP_004007747.2 | 5 - 6 | 713 | 70 | |
XM_027968431.1 | XP_027824232.1 | 4 - 5 | 837 | 70 | ||
XM_027968432.1 | XP_027824233.1 | 5 - 6 | 702 | 70 |