Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM41A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015092511.2 | XP_014947997.2 | 4 - 5 | 622 | 91 | |
XM_027957537.1 | XP_027813338.1 | 4 - 5 | 837 | 91 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Ovis aries |