Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NLRP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012185551.3 | XP_012040941.2 | 11 - 12 | 3414 | 101 | |
XM_027975093.1 | XP_027830894.1 | 10 - 11 | 3222 | 101 | ||
XM_027975095.1 | XP_027830896.1 | 11 - 12 | 3266 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |