Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MIS12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004012586.4 | XP_004012635.2 | 2 | 1614 | 82 | |
XM_027975092.1 | XP_027830893.1 | 2 | 1900 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Ovis aries |