Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAQR7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027965493.1 | XP_027821294.1 | 3 | 1226 | 75 | |
XM_027965494.1 | XP_027821295.1 | 4 | 1111 | 75 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor 3' Most Recommended Assay Ovis aries |