Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRDM4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001171358.1 | NP_001164829.1 | 8 - 9 | 1940 | 77 | |
XM_008257056.2 | XP_008255278.1 | 9 - 10 | 2005 | 77 | ||
XM_008257057.2 | XP_008255279.1 | 10 - 11 | 2242 | 77 | ||
XM_008257059.2 | XP_008255281.1 | 8 - 9 | 1859 | 77 | ||
XM_017341624.1 | XP_017197113.1 | 9 - 10 | 2265 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Oryctolagus cuniculus Probe spans exons |