Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AAR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002710823.3 | XP_002710869.1 | 2 - 3 | 2595 | 72 | |
XM_008256039.2 | XP_008254261.1 | 2 - 3 | 913 | 72 | ||
XM_017341642.1 | XP_017197131.1 | 1 - 2 | 758 | 72 | ||
GenBank mRNA | AF078806.2 | - | 2 - 3 | 672 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Oryctolagus cuniculus Probe spans exons |