Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NGB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082133.2 | NP_001075602.1 | 3 - 4 | 327 | 95 | |
XM_008271894.2 | XP_008270116.1 | 3 - 4 | 838 | 95 | ||
GenBank mRNA | AJ781214.1 | - | 3 - 4 | 331 | 95 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Oryctolagus cuniculus Probe spans exons |