Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRPC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082779.1 | NP_001076248.1 | 10 - 11 | 3008 | 108 | |
GenBank mRNA | AJ006203.1 | - | 10 - 11 | 3008 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Oryctolagus cuniculus Probe spans exons |