Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADRA1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082380.1 | NP_001075849.1 | 2 | 1046 | 106 | |
GenBank mRNA | AF157505.1 | - | 2 | 1046 | 106 | |
AF157506.1 | - | 2 | 1046 | 106 | ||
U81982.1 | - | 2 | 1046 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Detect Genomic DNA Oryctolagus cuniculus Both primers and probe map within a single exon |