Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FTO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199368.1 | NP_001186297.1 | 2 - 3 | 140 | 81 | |
XM_017342183.1 | XP_017197672.1 | 2 - 3 | 169 | 81 | ||
XM_017342184.1 | XP_017197673.1 | 2 - 3 | 169 | 81 | ||
GenBank mRNA | FJ429175.2 | - | 2 - 3 | 140 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Oryctolagus cuniculus Probe spans exons |