Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UCP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001171077.1 | NP_001164548.1 | 4 - 5 | 748 | 98 | |
GenBank mRNA | X14696.1 | - | 748 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Oryctolagus cuniculus Probe spans exons |