Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001171129.1 | NP_001164600.1 | 1 | 1454 | 79 | |
XM_008264050.2 | XP_008262272.2 | 1 | 1454 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Oryctolagus cuniculus Both primers and probe map within a single exon |