Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSMB4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001171305.1 | NP_001164776.1 | 2 - 3 | 342 | 75 | |
XM_008264042.2 | XP_008262264.1 | 2 - 3 | 346 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Oryctolagus cuniculus Probe spans exons |