Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EMD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001171372.1 | NP_001164843.1 | 80 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Oryctolagus cuniculus |