Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BLOC1S2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266915.1 | NP_001253844.1 | 404 | 69 | ||
XM_015147797.2 | XP_015003283.1 | 709 | 69 | |||
XM_015147798.2 | XP_015003284.1 | 1847 | 69 | |||
GenBank mRNA | JU319720.1 | - | 485 | 69 | ||
JV600271.1 | - | 404 | 69 | |||
JV712459.1 | - | 489 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Macaca mulatta |