Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNIH4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260666.1 | NP_001247595.1 | 283 | 96 | ||
GenBank mRNA | JU321164.1 | - | 282 | 96 | ||
JU472037.1 | - | 283 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Macaca mulatta |