Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KDM7A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015135046.2 | XP_014990532.1 | 978 | 101 | ||
GenBank mRNA | JU326136.1 | - | 867 | 101 | ||
JV724219.1 | - | 779 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |