Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRY2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015114399.2 | XP_014969885.1 | 409 | 58 | ||
GenBank mRNA | JU327095.1 | - | 283 | 58 | ||
JV495396.1 | - | 269 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Macaca mulatta |