Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCDH19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001265688.1 | NP_001252617.1 | 2634 | 81 | ||
XM_015127782.2 | XP_014983268.1 | 3115 | 81 | |||
XM_015127783.2 | XP_014983269.1 | 3115 | 81 | |||
XM_028841759.1 | XP_028697592.1 | 3256 | 81 | |||
GenBank mRNA | JV565155.1 | - | 2493 | 81 | ||
JV565156.1 | - | 2634 | 81 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Macaca mulatta |