Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STARD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260934.1 | NP_001247863.1 | 404 | 125 | ||
XM_015140561.2 | XP_014996047.1 | 605 | 125 | |||
XM_015140562.2 | XP_014996048.1 | 614 | 125 | |||
GenBank mRNA | JU332969.1 | - | 404 | 125 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |