Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SUV39H1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261327.1 | NP_001248256.1 | 1157 | 61 | ||
XM_015127316.2 | XP_014982802.1 | 1431 | 61 | |||
GenBank mRNA | JU328382.1 | - | 324 | 61 | ||
JU473137.1 | - | 1157 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Macaca mulatta |